Learn More
Abeba™ Anatom 2125 Shoes
Product Code. 15687817
Change view
Click to view available options
Size:
36
37
38
39
40
41
42
43
44
45
46
47
48
Unit Size:
1 pair
Product Code. Size unitSize
15687817 45 1 pair
15697807 36 1 pair
15607817 37 1 pair
15617817 38 1 pair
15627817 39 1 pair
15637817 40 1 pair
15647817 41 1 pair
15657817 42 1 pair
15667817 43 1 pair
15677817 44 1 pair
15697817 46 1 pair
15607827 47 1 pair
15617827 48 1 pair
13 options
Denne vare kan ikke returneres. Se returpolicy
Artikelnummer. 15687817
Brand: Abeba™ 2125/45
Kun null tilbage
Læg i indkøbskurv
Læg i indkøbskurv
Denne vare kan ikke returneres. Se returpolicy
Resperation absorbing special lining with silver ions. Steel toe-cap.
  • CE EN ISO 20345:2011 SB A, E SRC
  • Microfiber black
  • Steel toe cap
  • Shock absorbant PU midsole
  • Slip resistance PU outsole sole
  • Replaceable acc Wave insole (article no. 3582)
  • Sanitized treated
  • Antistatic
  • Heel strap non-foldable, adjustable
  • Breathable lining with Silver ions
  • Instep area adjustable, with elastic band
  • Suitable for heel and foot lift
Spezifikation
Certifications/Compliance CE EN ISO 20345:2011 SB A, E SRC
Color Black
Material Microfiber
Size 45
Sole Features Slip-resistant
Sole Material Polyurethane
Style 2125
Toe Material Steel