Learn More
Motic™ Analyzer, AE2000MET Series
Description
- Improve image quality obtained with birefringent materials when compared to other techniques such as darkfield and brightfield illumination, differential interference contrast, phase contrast, Hoffman modulation contrast, and fluorescence.
- Useful for both quantitative and qualitative studies targeted at a wide range of anisotropic specimens.
- Polarized light microscopy is capable of providing information on absorption color and optical path boundaries between minerals of differing refractive indices, in a manner similar to brightfield illumination, but the technique can also distinguish between isotropic and anisotropic substances.
- Contrast-enhancing technique exploits the optical properties specific to anisotropy and reveals detailed information concerning the structure and composition of materials that are invaluable for identification and diagnostic purposes.
Specifications
Specifications
| Type | Analyzer |
| For Use With (Equipment) | AE2000MET Series Microscopes |