Learn More
Thermo Scientific™ Analytical Membrane Bands
GREENER_CHOICE
Product Code. 17816078
Change view
Click to view available options
Color:
Black
White
Pore Size:
0.2 μm
0.45 μm
0.8 μm
Unit Size:
Pack of 600
Product Code. Color Pore Size unitSize
17816078 Black 0.45 μm Pack of 600
17806078 White 0.2 μm Pack of 600
17846078 White 0.8 μm Pack of 600
17836078 White 0.45 μm Pack of 600
17896068 Black 0.2 μm Pack of 600
17826078 Black 0.8 μm Pack of 600
6 options
Product Code. 17816078
Brand: Thermo Scientific™ DS06000245
Only null left
Add to basket
Add to basket
Thermo Scientific Analytical Membrane Bands are perfect for both microbial QA and QC applications where membrane dispensers are used. Product is compatible with most common dispensers on the market, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
Thermo Scientific™ Analytical Membrane Bands are perfect for microbial QA/QC applications where membrane dispensers are used. Compatible with most common dispensers on the market, including MilliporeSigma™ and Sartorius™, giving added flexibility and security of supply. Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading. Bands are numbered to indicate how many membranes remain to be dispensed. Membranes are gridded to improve accuracy and save time. Product has been validated to the highest standards.
Thermo Scientific™ Analytical Membrane Bands are perfect for both microbial quality assurance (QA) and quality control (QC) applications where membrane dispensers are used.
  • Product is compatible with most common dispenser platforms on the market, including MilliporeSigma™ and Sartorius™, giving additional flexibility and security of supply from the leading lab and life sciences supplier.
  • 47mm disks. Mixed cellulose esters (MCE). Available in both white and Black.
  • Each case of pre-sterilized Analytical Membrane Bands contains four boxes of 150 membranes per pack (600/case total).
  • Boxes feature two marked orientations and tear-away perforations with a thumb-tab for quick and easy loading.
  • Bands are numbered to indicate how many membranes remain to be dispensed.
  • Membranes are gridded to improve accuracy and save time.
  • Product has been validated to the highest standards, including ISO 7704 and ASTM F838.
TRUSTED_SUSTAINABILITY
Specifications
Color Black
Material (Membrane) MCE
Pore Size 0.45 μm
Quantity 600/Cs.
Diameter (Metric) Disc 47 mm