Learn More
IKA™ Analytical Batch Mill
Product Code. 13450607 Shop All IKA Products
Change view
Click to view available options
Model:
A 10 Basic
A 11 Basic
Capacity (Metric):
50 mL
80 mL
Hardness:
5 Mohs
6 Mohs
Dimensions (L x W x H):
85 x 85 x 240 mm
145 x 130 x 250 mm
Unit Size:
Each
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Model Capacity (Metric) Hardness Dimensions (L x W x H) Unit Size
13450607 A 10 Basic 50 mL 5 Mohs 145 x 130 x 250 mm Each
10479412 A 11 Basic 80 mL 6 Mohs 85 x 85 x 240 mm Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 13450607 Supplier IKA™ Supplier No. 0004020700
  • ONLINE EXCLUSIVE PRICE
    28135.00 NOK
    26080.62 NOK / Pack of 1
    Save 2054.38 NOK
    7% Off
View all promotions
In Stock
Grinds hard, brittle, soft and fibrous materials. IKA™ Analytical Batch Mill allows samples to be embrittled directly in the grinding chamber, tough, oily and aqueous samples can also be ground. Available models include the newly redesigned A 10 Basic and the A 11 Basic.
Batch mill for two different grinding procedures. Impact grinding of hard, brittle or non-elastic grinding materials with high grade stainless steel beater. Beater (supplied) can be used for a Mohs hardness up to six. Cutting grinding for pulverising soft, fibrous materials with a cutting blade (must be ordered separately).
  • Moist and gluey materials can be pulverised by adding water
  • Grinding chamber (included with mill) made of ETFE (PTFE, glass fibre reinforced) with stainless steel inlet (A1S1 3/6L), with a useful volume of 80mL
  • Can be used for the embrittlement of grinding materials with liquid nitrogen in the grinding chamber
  • 250mL grinding chamber is available
TRUSTED_SUSTAINABILITY
Specifications
Product Type Analytical Batch Mill
Capacity (Metric) 50 mL
Certifications/Compliance DIN EN 60529 IP 41
Dimensions (L x W x H) 145 x 130 x 250 mm
Electrical Requirements 100/220 V, 50/60 Hz, 300 W
Hardness 5 Mohs
Height (Metric) 250 mm
Material Stainless Steel
Model A 10 Basic
Particle Size 6 mm
Voltage 220/240 V
Watts 300 W
Weight (Metric) 2.9 kg
Width (Metric) 130 mm
Frequency 50/60 Hz