Learn More
BINDER™ Analog Output
Shop All BINDER Products
Click to view available options
For Use With (Equipment):
KB 65/130/260/470/720/1060
KB PRO 130/260/470
KB PRO Series
KBF Series
Unit Size:
Each
Description
- Analog output
- Not adjustable
- For temperature values or temperature and humidity values
- 4 to 20 mA
Specifications
Specifications
| For Use With (Equipment) | KBF Series |
| Material | Copper |
| Type | Analog Output |
| Features | For Temperature and Humidity Values |