Learn More
Interscience™ Anabac Deodorizing Capsules
Product Code. 16825923
Change view
Click to view available options
Scent:
Apple
Citrus
Fresh Scent
Color:
Clear
Green, White
Yellow
Appearance:
Clear Amber
Yellow
Sterility:
Non-sterile
Sterile
Unit Size:
Each
Pack of 100
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Scent Color Appearance Sterility Unit Size
16825923 Citrus Yellow Yellow Non-sterile Pack of 100
10422084 Fresh Scent Clear Yellow Sterile Each
15388927 Apple Green, White Clear Amber Sterile Pack of 100
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16825923 Supplier Interscience™ Supplier No. 320300
  • ONLINE EXCLUSIVE PRICE
    1205.00 NOK
    1171.04 NOK / Pack of 100
    Save 33.96 NOK
    2% Off
View all promotions
 Restricted Item
Currently Unavailable: We have no information when this product will be available.
View alternative products
Placed in an autoclave, Anabac capsules provide a discreet way to perfume the laboratory.
  • Capsule dissolves during an autoclaving destruction cycle
  • Capsule melts completely without leaving residue
  • Eliminates unpleasant odors that can emerge from the autoclave
Specifications
Product Type Autoclave Capsule
Bristle Material Plastic
Color-coded Yes
Form Capsules
Handle Material Plastic
Handle Type Cap
Scent Citrus
Sterility Non-sterile
Appearance Yellow
Autoclavable Autoclavable
Color Yellow
Disposable Single-use
Filtered Non-filtered
For Use With (Application) Refreshes and Perfumes Air Where Autoclave Is Located
For Use With (Equipment) Autoclave
Quantity 100 Capsules/Pk.