Learn More
Amyl Nitrite, TRC
Specifications
Specifications
| Chemical Name or Material | Amyl Nitrite |
| CAS | 463-04-7 |
| Molecular Formula | C5 H11 N O2 |
| Quantity | 1 mL |
| Synonym | 1-Pentyl Nitrite, Nitramyl, n-Amyl Nitrite, n-Pentyl Nitrite |
| InChI Formula | InChI=1S/C5H11NO2/c1-2-3-4-5-8-6-7/h2-5H2,1H3 |
| SMILES | CCCCCON=O |
| IUPAC Name | pentyl nitrite |
| Molecular Weight (g/mol) | 117.15 |
| Formula Weight | 117.079 |