Learn More
Amsacrine hydrochloride, MedChemExpress
Product Code. 16465748
Encompass_Preferred
Change view
Click to view available options
Quantity:
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16465748 10 mg 10mg
16461907 50 mg 50mg
16446226 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 16465748 Supplier Medchem Express Supplier No. HY13551A
 Restricted Item
Estimated Shipment: 27-11-2026
MedChemExpress Amsacrine hydrochloride (m-AMSA hydrochloride; acridinyl anisidide hydrochloride) is an inhibitor of topoisomerase II, and acts as an antineoplastic agent which can intercalates into the DNA of tumor cells.
Amsacrine hydrochloride (m-AMSA hydrochloride
Specifications
Chemical Name or Material Amsacrine hydrochloride
CAS 54301-15-4
Color Orange
Purity Grade Notes Research
Molecular Formula C21H20ClN3O3S
Quantity 10 mg
Synonym m-AMSA hydrochloride acridinyl anisidide hydrochloride
Solubility Information DMSO : 62.5 mg/mL (145.38 mM; Need ultrasonic)
SMILES CS(=O)(NC1=CC=C(NC2=C(C=CC=C3)C3=NC4=CC=CC=C42)C(OC)=C1)=O.[H]Cl
Molecular Weight (g/mol) 429.92
Formula Weight 429.92
Percent Purity 98.98%
Grade Research
Physical Form Solid
For Use With (Application) Cancer-programmed cell death