Learn More
AMPK beta 2 Mouse anti-Human, Mouse, Rat, Clone: 2D5C1, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech 66579-1-IG-150UL
 View more versions of this product
Product Code. 16838585
  • 4836.10 NOK / 150µL
Description
AMPK Beta 2 is a regulatory subunit of the AMP-activated protein kinase (AMPK). AMPK is a heterotrimer consisting of an alpha catalytic subunit, and non-catalytic beta and gamma subunits. AMPK is an important energy-sensing enzyme that monitors cellular energy status. In response to cellular metabolic stresses, AMPK is activated, and thus phosphorylates and inactivates acetyl-CoA carboxylase (ACC) and beta-hydroxy beta-methylglutaryl-CoA reductase (HMGCR), key enzymes involved in regulating de novo biosynthesis of fatty acid and cholesterol. This subunit may be a positive regulator of AMPK activity. The myristoylation and phosphorylation of this subunit have been shown to affect the enzyme activity and cellular localization of AMPK. This subunit may also serve as an adaptor molecule mediating the association of the AMPK complex.
Specifications
Specifications
AMPK beta 2
Monoclonal
1 mg/mL
PBS with 50% glycerol and 0.1% sodium azide; pH 7.3
O43741, Q6PAM0, Q9QZH4
PRKAB2
AMPK beta 2 Fusion Protein Ag5806
150 μL
Primary
Human, Mouse, Rat
Antibody
IgG1
Western Blot
2D5C1
Unconjugated
PRKAB2
AMPK subunit beta 2, PRKAB2
Mouse
Protein G
RUO
108097, 5565, 64562
-20°C