missing translation for 'onlineSavingsMsg'
Learn More

TOKU-E™ Ampicillin ReadyMade™ Solution

Product Code. p-200102278
Encompass_Preferred
Click to view available options
Quantity:
10 mL
5 x 10 mL
Unit Size:
10mL
50mL
This item is not returnable. View return policy

Product Code. 30044039

Brand: TOKU-E™ A22810ML

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Ampicillin ReadyMade™ Solution is provided as a filter-sterilized solution of ampicillin anhydrous formulated in DMSO.

  • CAS Number: 69-53-4
  • Molecular Formula: C16H19N3O4S (ampicillin anhydrous)
  • Molecular Weight: 349.40 g/mol
  • Storage Conditions: -20°C
  • Form: Liquid
  • Source: Semi-synthetic
  • Spectrum: Ampicillin is broad-spectrum, effective against both Gram-positive and Gram-negative bacteria including Gram-negative non-extended spectrum β-lactamase (ESBL) bacteria including Staphylococcus and Streptococcus species and medically important enteric pathogens such as Shigella and Salmonella. Interestingly, Ampicillin has been found to be effective against certain β-lactam sensitive vancomycin-resistant Enterococcus (VRE); a glycopeptide antibiotic resistant 'superbug'. Resistance to Ampicillin is routinely used as a selectable marker to confirm successful cell transformation
  • Mechanism of Action: Like all β-lactams, Ampicillin interferes with penicillin binding protein (PBP) activity, otherwise involved in the final phase of peptidoglycan synthesis. PBPs are enzymes which catalyze a pentaglycine crosslink between alanine and lysine residues. Without a pentaglycine crosslink, the integrity of the cell wall is severely compromised ultimately leading to cell lysis

Specifications

CAS 69-53-4
Certifications/Compliance Purity >98.0%
Chemical Name or Material Ampicillin
Color Opaque white to opaque yellow
Content And Storage -20°C
Description Ampicillin Readymade™ Solution 100 mg/mL in DMSO, Sterile Filtered
Concentration 100 to 105 mg/mL
Form Liquid
Product Type Antibiotic
Sterility Sterile
Sterilization Method Sterile-filtered
With Additives None
Without Additives None
Formulation Liquid, in Dimethyl Sulfoxide
For Use With (Application) Microbiology, Eukaryotic Cell Culture Applications
Product Line ReadyMade™ Solutions
Purity or Quality Grade >98.0%
Quantity 10 mL
Shipping Condition Ice Pack
Source Semi-synthetic
Show More Show Less
Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.