missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Amphiregulin Mouse anti-Human, Rat, Clone: 1A1G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech 66433-1-IG-20UL
This item is not returnable.
View return policy
Description
The protein encoded by this gene is a member of the epidermal growth factor family. It is an autocrine growth factor as well as a mitogen for astrocytes, Schwann cells, and fibroblasts. It is related to epidermal growth factor (EGF) and transforming growth factor alpha (TGF-alpha). This protein interacts with the EGF/TGF-alpha receptor to promote the growth of normal epithelial cells and inhibits the growth of certain aggressive carcinoma cell lines. This encoded protein is associated with a psoriasis-like skin phenotype.Specifications
| Amphiregulin | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P15514, P24338 | |
| Areg | |
| Amphiregulin Fusion Protein Ag8907 | |
| 20 μL | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG1 |
| Western Blot, Immunohistochemistry (Paraffin) | |
| 1A1G9 | |
| Unconjugated | |
| AREG | |
| amphiregulin, AR, AREG, AREGB, CRDGF, SDGF | |
| Mouse | |
| Protein A | |
| RUO | |
| 29183, 374 | |
| -20°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction