Learn More
AMPD, 0.2M buffer soln., pH 8.9, Thermo Scientific™
Product Code. p-7042109
Click to view available options
Quantity:
100 mL
250 mL
Unit Size:
100mL
250mL
Product Code. 15463979
Brand: Thermo Scientific Alfa Aesar J61377.AE
Request Bulk or Custom Format
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
This Thermo Scientific brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific.
TRUSTED_SUSTAINABILITY
Specifications
CAS 115-69-5
Color Colorless
Concentration 0.2 M
pH 8.9
Physical Form Liquid
Quantity 100 mL
Odor Odorless
Molecular Weight (g/mol) 105.137