Learn More
AMOZ-d5, TRC
Product Code. 30338375
Change view
Click to view available options
Quantity:
1 mg
Unit Size:
1mg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
30338375 1 mg 1mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
Product Code. 30338375 Supplier Toronto Research Chemicals Supplier No. TRCA6346021MG
 Restricted Item
Estimated Shipment: 12-11-2026
Regulatory: Restrictions may apply to the purchase of this product and we may need to request additional documentation from you
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Specifications
Chemical Name or Material 3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5)
CAS 1017793-94-0
Purity Grade Notes HPLC
Molecular Formula C8 2H5 H10 N3 O3
Quantity 1 mg
Synonym 3-Amino-5-morpholinomethyl-2-oxazolidinone D5 (AMOZ D5), 2-Oxazolidinone-4,4,5-d3, 3-amino-5-(4-morpholinylmethyl-d2)-
InChI Formula InChI=1S/C8H15N3O3/c9-11-6-7(14-8(11)12)5-10-1-3-13-4-2-10/h7H,1-6,9H2/i5D2,6D2,7D
SMILES [2H]C([2H])(N1CCOCC1)C2([2H])OC(=O)N(N)C2([2H])[2H]
IUPAC Name 3-amino-4,4,5-trideuterio-5-[dideuterio(morpholin-4-yl)methyl]-1,3-oxazolidin-2-one
Molecular Weight (g/mol) 206.25
Formula Weight 206.1427
Percent Purity >95