Learn More
Ammonium Thiocyanate, 0.1M, Ph. Eur., Reagecon™
Shop All Reagecon Products
Click to view available options
Quantity:
1 L
Unit Size:
1 L
Specifications
Specifications
| Quantity | 1 L |
| Grade | Ph. Eur. |
| Concentration or Composition (by Analyte or Components) | 0.1M |