Learn More
Reagecon™ Ammonium sulfate solution 2M/L electrode filling soln,dble junction bridge sol Reagecon
Product Code. 11533172 Shop All Reagecon Products
Product Code. 11533172
Brand: Reagecon™ EFS2AS
compliance-icons
Product Identifier
  • ELECTRODE FILLING SOL. DOUBLE JUNCTION BRIDGE SOL. 2M AMMONIUM SULPHATE (NH
Signal Word
  • Warning
Hazard Category
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity Category 3
Hazard Statement
  • H315-Causes skin irritation.
  • H319-Causes serious eye irritation.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
  • MIXTURE LIST-Contains : ammonium sulphate