missing translation for 'onlineSavingsMsg'
Learn More

Ammonia Standard Solution, Ion HIQU, Chem-Lab

Product Code. p-8014391
Click to view available options
Quantity:
100 mL
500 mL
Unit Size:
100mL
500mL
This item is not returnable. View return policy

Product Code. 11969012

Brand: Chem-Lab CL01.0111.0100

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Contains 1000μg/mL of NH4+. Ammonia Standard Solution, Ion HIQU, Chem-Lab is a single component solution with 2.97g NH4Cl per 1L of H2O.

Ammonium standard solution is used as an analytical/elemental standard and in ion chromatography. It is used in ammonia determination and to calibrate equipment with ammonia sensors.

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
PubChem CID 25517
ChEBI CHEBI:31206
IUPAC Name amine hydrochloride

Specifications

Odor Odorless
Density 1.00g/mL
Physical Form Liquid
Concentration 2.97g NH4Cl per 1L of H2O
Color Colorless
pH ±5
Solubility Soluble
SMILES N.Cl
Quantity 100 mL
Formula Weight 53.49g/mol
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.