missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Phosphate Dibasic, Reagent Grade, ≥98%, Honeywell™

Product Code. 15607660 Shop All Honeywell Research Chemicals Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
2.5 kg
Packaging:
Poly Bottle
Unit Size:
100g
1kg
2.5kg
500g
Product Code. Quantity Packaging unitSize
15607660 500 g Poly Bottle 500g
15677650 100 g Poly Bottle 100g
15687650 1 kg Poly Bottle 1kg
15697650 2.5 kg Poly Bottle 2.5kg
4 options
This item is not returnable. View return policy

Product Code. 15607660

Brand: Honeywell Research Chemicals A5764500G

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Reagent Grade,≥98.0%

  • ≥95% for general laboratory use.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7783-28-0
Molecular Formula H9N2O4P
Molecular Weight (g/mol) 132.06
MDL Number MFCD00010891
InChI Key MNNHAPBLZZVQHP-UHFFFAOYSA-N
Synonym diammonium phosphate, diammonium hydrogenphosphate, diammonium hydrogen phosphate, phosphoric acid, diammonium salt, fyrex, pelor, akoustan a, diammonium orthophosphate, diammonium acid phosphate, dibasic ammonium phosphate
PubChem CID 24540
ChEBI CHEBI:63051
SMILES N.N.OP(O)(O)=O

Specifications

Melting Point 155°C
Color White
pH 7.5 to 9 (at 25°C)
Quantity 500 g
Linear Formula (NH4)2HPO4
UN Number NONH for all modes of transport
IUPAC Name diazanium;hydrogen phosphate
Formula Weight 132.06g/mol
Percent Purity ≥98%
Odor Odorless
Packaging Poly Bottle
Appearance Crystalline
Density 1.62g/cm3 at 20°C
Chemical Name or Material Ammonium phosphate dibasic
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.