Learn More
Ammonium Peroxodisulfate, MilliporeSigma™
Product Code. 15844050
Change view
Click to view available options
Quantity:
500 g
5 kg
Packaging:
Plastic Bottle
Unit Size:
500g
5kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15844050 5 kg Plastic Bottle 5kg
15834050 500 g Plastic Bottle 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Product Code. 15844050 Supplier Supelco Supplier No. 1012015000
 Restricted Item
Estimated Shipment: 25-11-2026
For analysis EMSURE™ ACS, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7727-54-0
Molecular Formula H8N2O8S2
Molecular Weight (g/mol) 228.19
MDL Number MFCD00003390
InChI Key ROOXNKNUYICQNP-UHFFFAOYSA-N
Synonym ammonium persulfate, ammonium peroxydisulfate, diammonium peroxydisulfate, diammonium persulfate, ammoniumpersulfate, diammonium peroxodisulphate, diammonium peroxydisulphate, unii-22qf6l357f, ccris 1430, persulfate d'ammonium french
PubChem CID 62648
IUPAC Name diammonium [(sulfonatoperoxy)sulfonyl]oxidanide
SMILES [NH4+].[NH4+].[O-]S(=O)(=O)OOS([O-])(=O)=O
Specifications
Physical Form Solid
Quantity 5 kg
UN Number UN1444
Solubility Information Soluble in water: 620g/L at 20°C
Formula Weight 228.19
Percent Purity ≥98.0%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 1.98g/cm3 at 20°C
Chemical Name or Material Ammonium peroxodisulfate