missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Oxalate Monohydrate, Certified AR for analysis, Fisher Chemical™

Product Code. 10213230 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
1 kg
Unit Size:
100g
1kg
500g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10213230 1 kg 1kg
10438590 100 g 100g
10705901 500 g 500g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
This item is not returnable. View return policy
Product Code. 10213230 Supplier Fisher Chemical Supplier No. A/6040/60
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Ammonium Oxalate Monohydrate, Certified AR for analysis

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 6009-70-7
Molecular Formula C2H10N2O5
Molecular Weight (g/mol) 142.11
MDL Number 149694
InChI Key MSMNVXKYCPHLLN-UHFFFAOYSA-N
IUPAC Name oxalic acid diamine hydrate
SMILES N.N.O.OC(=O)C(O)=O

Specifications

Melting Point 70°C
Color White
pH 6.4
Physical Form Solid
Quantity 1 kg
Formula Weight 142.11g/mol
Packaging Plastic powder jar
compliance-icons
Product Identifier
  • Ammonium oxalate monohydrate
Signal Word
  • Warning
Hazard Category
  • Acute toxicity Category 4
  • Serious eye damage/eye irritation Category 2
Hazard Statement
  • H312-Harmful in contact with skin.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P312-Call a POISON CENTER/doctor/if you feel unwell.
Titolo del prodotto
Selezionare un problema

Facendo clic su Invia, l'utente riconosce che potrebbe essere contattato da Fisher Scientific in merito al feedback fornito in questo modulo. Non condivideremo le vostre informazioni per altri scopi. Tutte le informazioni di contatto fornite saranno conservate in conformità con la nostra Politica sulla privacy. Informativa sulla privacy.