Learn More
Ammonium Nitrate, MilliporeSigma™
Product Code. 15896960
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
Packaging:
Plastic Bottle
Unit Size:
1kg
500g
5kg
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15896960 1 kg Plastic Bottle 1kg
15874040 500 g Plastic Bottle 500g
15806970 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
Product Code. 15896960 Supplier Supelco Supplier No. 1011881000
 Restricted Item
Estimated Shipment: 26-11-2026
Regulatory: This product contains an explosive precursor according to Regulation (EC) No 1148/2019, all customers must complete a declaration before delivery.
For analysis EMSURE™ ACS
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 6484-52-2
Molecular Formula H4N2O3
Molecular Weight (g/mol) 80.04
MDL Number MFCD00011425
InChI Key DVARTQFDIMZBAA-UHFFFAOYSA-O
Synonym ammonium nitrate, nitram, ammonium nitricum, ammonium saltpeter, nitrate of ammonia, nitric acid ammonium salt, nitrato amonico, herco prills, german saltpeter, merco prills
PubChem CID 22985
ChEBI CHEBI:63038
IUPAC Name ammonium nitrate
SMILES [NH4+].[O-][N+]([O-])=O
Specifications
Boiling Point 210°C
Physical Form Solid
Quantity 1 kg
UN Number UN1942
Solubility Information Soluble in water: 1920g/L at 20°C
Formula Weight 80.04
Percent Purity 95%
Grade ACS
Odor Odorless
Packaging Plastic Bottle
Density 1.72g/cm3 at 20°C
Chemical Name or Material Ammonium nitrate