Learn More
Ammonium iron(III) sulfate dodecahydrate, MilliporeSigma™
Product Code. 15834370
Change view
Click to view available options
Quantity:
500 g
1 kg
5 kg
50 kg
Packaging:
Fibre carton
Plastic Bottle
Unit Size:
1kg
500g
50kg
5kg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Packaging Unit Size
15834370 50 kg Fibre carton 50kg
15804370 500 g Plastic Bottle 500g
15814370 1 kg Plastic Bottle 1kg
15824370 5 kg Plastic Bottle 5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 15834370 Supplier Supelco Supplier No. 1037769050
 Restricted Item
Estimated Shipment: 25-11-2026
For analysis EMSURE™ ACS, ISO, Reag. Ph Eur
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 7783-83-7
Molecular Formula FeH28NO20S2
Molecular Weight (g/mol) 482.176
InChI Key LCPUDZUWZDSKMX-UHFFFAOYSA-K
Synonym ammonium ferric sulfate, ferric ammonium sulfate dodecahydrate, iron ammonium sulfate dodecahydrate, sulfuric acid, ammonium iron 3+ salt 2:1:1 , dodecahydrate, ferric ammonium sulfate, acs, azanium iron 3+ disulfate dodecahydrate, ferric iron ammonium dodecahydrate disulfate, azanium iron +3 cation disulfate dodecahydrate, ammonium iron iii sulfate dodecahydrate, acs 100g
PubChem CID 197097
IUPAC Name azanium;iron(3+);disulfate;dodecahydrate
SMILES [NH4+].O.O.O.O.O.O.O.O.O.O.O.O.[O-]S(=O)(=O)[O-].[O-]S(=O)(=O)[O-].[Fe+3]
Specifications
Physical Form Solid
Quantity 50 kg
Solubility Information Soluble in water: 1240g/L at 25°C
Formula Weight 482.19
Grade ACS
Odor Odorless
Packaging Fibre carton
Chemical Name or Material Ammonium iron(III) sulfate dodecahydrate