Learn More
Ammonium Dihydrogen Phosphate, Supelco™
Specifications
Specifications
| Melting Point | 190°C |
| CAS | 7722-76-1 |
| Quantity | 50 g |
| Molecular Formula | (NH4)H2PO4 |
| MDL Number | MFCD00003396 |
| Synonym | Ammonium dihydrogen phosphate, Ammonium biphosphate, Ammonium phosphate monobasic, Primary ammonium phosphate, Monoammonium orthophosphate, Ammonium dihydrogen phosphate |
| Molecular Weight (g/mol) | 115.03 |
| Grade | Research |
| Density | 1.80 g/cm3 at 20°C |
| Chemical Name or Material | Ammonium Dihydrogen Phosphate |