missing translation for 'onlineSavingsMsg'
Learn More

Ammonium Chloride, ACS Reagent, Honeywell Fluka™

Product Code. 15641960 Shop All Honeywell Fluka Products
Change view
Click to view available options
Quantity:
25 g
500 g
1 kg
2.5 kg
4 x 2.5 kg
Unit Size:
1kg
2.5kg
25g
500g
Pack of 4
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15641960 2.5 kg 2.5kg
15651960 25 g 25g
15671960 500 g 500g
15631960 1 kg 1kg
15661960 4 x 2.5 kg Pack of 4
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
This item is not returnable. View return policy
Product Code. 15641960 Supplier Honeywell-Fluka Supplier No. 2133302.5KG
Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

ACS Reagent,≥99.5%

  • Leading the Industry For Over 65 Years
  • Honeywell now delivers Fluka™ premium grade inorganic reagents worldwide – with consistency, purity, and accuracy assured
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 12125-02-9
Molecular Formula ClH4N
Molecular Weight (g/mol) 53.49
MDL Number MFCD00011420
InChI Key NLXLAEXVIDQMFP-UHFFFAOYSA-N
Synonym ammonium chloride, salmiac, sal ammoniac, ammonium muriate, ammoniumchlorid, ammoneric, darammon, amchlor, ammonium chloride nh4 cl, ammoniumklorid
PubChem CID 25517
ChEBI CHEBI:31206

Specifications

Quantity 2.5 kg
Linear Formula NH4Cl
Beilstein 4371014
SMILES N.Cl
IUPAC Name azanium;chloride
Formula Weight 53.49g/mol
Percent Purity ≥99.5%
Grade ACS Reagent
Residue after Ignition ≤0.01%
Vapor Pressure 1mmHg (160.4°C)
Density Vapor Density: 1.9 (vs air)
Chemical Name or Material Ammonium Chloride
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.