missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Ammonium Chloride, ACS Reagent Grade, Scharlab
Click to view available options
Quantity:
500 g
1 kg
Unit Size:
1kg
500g
Description
- Synonyms: Salt ammoniac
- Molecular Formula: NH4Cl
- Molar Mass: 53.49g/mol
- CAS: 12125-02-9
- EIN ECS No.: 235-186-4
Specifications
Specifications
| Melting Point | 335°C (decomposition) |
| pH | 4.5–5.5 (5%, water, at 25°C) |
| Quantity | 1 kg |
| Formula Weight | 53.49g/mol |
| Percent Purity | 99.5% min (argentometric) |
| Grade | ACS Reagent |
| Ignition Point | >400°C |
| Loss on Drying | 1% max (105°C) |
| Residue after Ignition | 0.01% max (as SO4) |
| Appearance | Passes test |
| Show More |
Safety and Handling
Product Identifier
- Ammonium chloride, reagent grade, ACS, ISO, Reag. Ph Eur
- Warning
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- H302-Harmful if swallowed.
- H319-Causes serious eye irritation.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P264-Wash thoroughly after handling.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P301+P312-IF SWALLOWED: Call a POISON CENTER or doctor/physician/ if you feel unwell.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P501-Dispose of contents/container to .
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction