Learn More
Ammonium and Cerium Sulphate solution, 0.01M EU Pharmacopoeia, Fisher Chemical™
Shop All Fisher Chemical Products
Click to view available options
Quantity:
1000 mL
Unit Size:
1000mL
Specifications
Specifications
| Chemical Name or Material | Ammonium and cerium sulphate solution |
| Packaging | Bottle |
| Concentration | 0.01 M |
| Quantity | 1000 mL |
| Grade | Eu. Ph. Reference |