Learn More
Ammonium and Cerium Nitrate solution, 0.1M EU Pharmacopoeia, Fisher Chemical™
BIOPHARMA
Product Code. 12900844 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
1000 mL
Packaging:
Bottle
Unit Size:
1000mL
Product Code. Quantity Packaging unitSize
12900844 1000 mL Bottle 1000mL
1 options
Product Code. 12900844
Brand: Fisher Chemical VL 002 1000
Pharmacopoeia reagents are intended for use in pharmacopoeia testing and monographs, produced under a GMP compliant ISO 9001 quality management system according to the relevant pharmacopoeia specification, using the pharmacopoeia reagent quality components and water meeting the relevant pharmacopoeia monograph requirement for Purified Water. Some pharmacopoeia standards are also accredited according to ISO 17025 and ISO Guide 43.
Specifications
Chemical Name or Material Ammonium and cerium nitrate solution
Quantity 1000 mL
Grade Eu. Ph. Reference
Concentration or Composition (by Analyte or Components) 0.1M
Packaging Bottle
compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonium and cerium nitrate 0.1M Volumetric Solution European Pharmacopoeia Ref: 3000100
Signal Word
  • Danger
Hazard Category
  • LONG-TERM AQUATIC HAZARD Chronic 2
  • Serious eye damage/eye irritation Category 1
  • Substances and mixtures corrosive to metals Category 1
  • Skin corrosion/irritation Category 1 C
  • Skin sensitiser Category 1
Hazard Statement
  • H290-May be corrosive to metals.
  • H314-Causes severe skin burns and eye damage.
  • H317-May cause an allergic skin reaction.
  • H318-Causes serious eye damage.
  • H411-Toxic to aquatic life with long lasting effects.
Precautionary Statement
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P273-Avoid release to the environment.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P390-Absorb spillage to prevent material damage.
  • P391-Collect spillage.
Supplemental information
  • MIXTURE LIST-Contains : sulphuric acid, ceric ammonium nitrate