Learn More
Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis, Fisher Chemical™
Product Code. p-7072849 Shop All Fisher Chemical Products
Click to view available options
Quantity:
250 mL
500 mL
1 L
Unit Size:
1 L
250mL
500mL
Product Code. 11944131
Brand: Fisher Chemical A470250
Request Bulk or Custom Format
Ammonia Solution 20-22%, Optima™, for Ultra Trace Elemental Analysis
TRUSTED_SUSTAINABILITY
Specifications
Color Colorless
CAS 1336-21-6
Molecular Formula H5NO
MDL Number 66650
Vapor Pressure 500 hPa 50
Vapor Density 0.59
pH 13.6
Quantity 250 mL
Physical Form Liquid
compliance-icons compliance-icons
Product Identifier
  • Ammonium hydroxide solution 20-22%, Optima for Ultra traces analysis
Signal Word
  • Danger
Hazard Category
  • Skin corrosion/irritation Category 1 B
  • Serious eye damage/eye irritation Category 1
  • Specific target organ toxicity Category 3
Hazard Statement
  • H314-Causes severe skin burns and eye damage.
  • H335-May cause respiratory irritation.
Precautionary Statement
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.