missing translation for 'onlineSavingsMsg'
Learn More

Ammonia, 2M in methanol

Product Code. 11406150
Change view
Click to view available options
Quantity:
100 mL
1000 mL
Unit Size:
1000mL
100mL
Product Code. Quantity unitSize
11406150 1000 mL 1000mL
11496140 100 mL 100mL
2 options
This item is not returnable. View return policy

Product Code. 11406150

Brand: Thermo Scientific Alfa Aesar H27080.AU

Request Bulk or Custom Format

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

CAS: 7664-41-7 | H3N | 17.03 g/mol

Ammonia, 2M in methanol is used as a solvent in organic reactions and as a mobile phase in thin layer chromatography.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 7664-41-7
Molecular Formula H3N
Molecular Weight (g/mol) 17.03
MDL Number MFCD00011418
InChI Key QGZKDVFQNNGYKY-UHFFFAOYSA-N
Synonym ammonia, ammonia gas, spirit of hartshorn, nitro-sil, ammoniakgas, ammonia anhydrous, anhydrous ammonia, ammonia, anhydrous, ammoniak, am-fol
PubChem CID 222
ChEBI CHEBI:16134
IUPAC Name azane
SMILES N

Specifications

Linear Formula NH3
UN Number UN2924
Concentration 2M in methanol
Density 0.787
Merck Index 14,492
Solubility Information Miscible with water.
Formula Weight 17.03
Quantity 1000 mL
Flash Point 6°C (43°F)
Sensitivity Hygroscopic
Chemical Name or Material Ammonia
Show More Show Less
compliance-icons compliance-icons compliance-icons compliance-icons
Product Identifier
  • Ammonia, 2M in methanol
Signal Word
  • Danger
Hazard Category
  • Acute toxicity Category 3
  • Serious eye damage/eye irritation Category 1
  • Flammable liquid Category 2
  • Skin corrosion/irritation Category 2
  • Specific target organ toxicity after single exposure Category 1
Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H301-Toxic if swallowed.
  • H311-Toxic in contact with skin.
  • H315-Causes skin irritation.
  • H318-Causes serious eye damage.
  • H331-Toxic if inhaled.
  • H370-Causes damage to organs.
Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/soap.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
  • MIXTURE LIST-Contains : Methanol, Ammonia

RUO – Research Use Only

Name des Produkts
Wählen Sie ein Thema

Indem Sie auf Absenden klicken, erklären Sie sich damit einverstanden, dass Fisher Scientific sich mit Ihnen in Verbindung setzen kann, um Ihr Feedback in diesem Formular zu bearbeiten. Wir werden Ihre Informationen nicht für andere Zwecke weitergeben. Alle bereitgestellten Kontaktinformationen werden in Übereinstimmung mit unserer Datenschutzrichtlinie aufbewahrt. Datenschutzrichtlinie.