Learn More
Aminophylline, TRC
Product Code. 30133644
Change view
Click to view available options
Quantity:
1 g
10 g
100 g
Unit Size:
100g
10g
1g
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
30133644 10 g 10g
30136265 100 g 100g
18011510 1 g 1g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Product Code. 30133644 Supplier Toronto Research Chemicals Supplier No. TRCA62775010G
This item requires a price update prior to being ordered.
Request a Price for this item here.
High-purity organic molecules and analytical standards, strategically delivered worldwide to empower innovation and commercial success.
TRUSTED_SUSTAINABILITY
Chemical Identifiers
CAS 317-34-0
Molecular Formula 2 C7 H8 N4 O2 . C2 H8 N2
Molecular Weight (g/mol) 420.43
Synonym Theophylline-Ethylenediamine, Aminophylline, 1H-Purine-2,6-dione, 3,9-dihydro-1,3-dimethyl-, compd. with 1,2-ethanediamine (2:1), 1H-Purine-2,6-dione, 3,7-dihydro-1,3-dimethyl-, compd. with 1,2-ethanediamine (2:1) (9CI), Theophylline, compd. with ethylenediamine (2:1) (8CI), 1,2-Ethanediamine, compd. with 3,7-dihydro-1,3-dimethyl-1H-purine-2,6-dione (1:2) (9CI), Ethylenediamine, compd. with theophylline (1:2) (8CI), Aminocardol, Aminodur, Aminofilina, Aminophyllin, Ammophyllin, Cardiofilina, Cardiomin, Cardophylin, Cardophyllin, Carena, Carine, Cidophylline, Diaphylline, Diophllin, Diophyllin, Diuxanthine, Dobo, Dura-Tab S.M. Aminophylline, Ethophylline, Euphyllin, Euphyllin CR, Euphyllin CR N 300, Euphylline, Eurphyllin, Euufillin, Genophyllin, Grifomin, Inophylline, Lasodex, Linampheta, Metaphyllin, Metaphylline, Methophylline, Minaphil, Minaphol, Miofilin, Neophyiline, Norofilina, Novophyllin, Novphyllin, Pecram, Peterphyllin, Phylcardin, Phyllindon, Phyllocontin, Pylcardin, Rectalad-aminophylline, Somophyllin, Somophyllin O, Stenovasan, Syntophyllin, Tefamin, Teofyllamin, Theodrox, Theolamine, Theolone, Theomin, Theophyldine, Theophyllamine, Theophyllaminum, Theophylline compd. with ethylenediamine, Vasofilina
IUPAC Name 1,3-dimethyl-7H-purine-2,6-dione;ethane-1,2-diamine
SMILES CN1C(=O)N(C)c2nc[nH]c2C1=O.CN3C(=O)N(C)c4nc[nH]c4C3=O.NCCN
Specifications
Quantity 10 g
InChI Formula InChI=1S/2C7H8N4O2.C2H8N2/c2*1-10-5-4(8-3-9-5)6(12)11(2)7(10)13;3-1-2-4/h2*3H,1-2H3,(H,8,9);1-4H2
Formula Weight 420.1982
Chemical Name or Material Theophylline-Ethylenediamine Anhydrous