missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Aminomethanephosphonic Acid-13C,15N, TRC
Click to view available options
Quantity:
10 mg
Unit Size:
10mg
Specifications
Specifications
| Chemical Name or Material | Aminomethanephosphonic Acid-13 C,15 N |
| Purity Grade Notes | HPLC |
| Molecular Formula | 13 C H6 15 N O3 P |
| Quantity | 10 mg |
| Synonym | P-(Aminomethyl)phosphonic Acid-13 C,15 N, AMPA-13 C,15 N, AMPA-13 C,15 N, Aminomethanephosphonic Acid-13 C,15 N, Aminomethylenephosphonic Acid-13 C,15 N, Phosphaglycine-13 C,15 N, Phosphaglycine-13 C,15 N, Phosphaglycine-13 C,15 N |
| InChI Formula | InChI=1 S/CH6NO3P/c2-1-6(3,4)5/h1-2H2,(H2,3,4,5)/i1+1,2+1 |
| SMILES | [15 NH2][13 CH2]P(=O)(O)O |
| IUPAC Name | (^{15}N)azanyl(1 ^{13}C)methylphosphonic acid |
| Molecular Weight (g/mol) | 113.0231 |
| Formula Weight | 113.0089 g/mol |
| Show More |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction