Learn More
Macherey-Nagel™ Amino Phases Polygosil™
Shop All Macherey-Nagel Products
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
10 μm
Unit Size:
100g
10g
Specifications
Specifications
| Particle Size | 10 μm |
| Pore Size | 60 Å |
| Technique | Liquid Chromatography |
| Quantity | 10 g |
| Type | Polygosil NH2 Phases |