Learn More
Macherey-Nagel™ Amino Phases Polygosil™
Product Code. 10608801 Shop All Macherey-Nagel Products
Change view
Click to view available options
Quantity:
10 g
100 g
Particle Size:
5 μm
10 μm
Unit Size:
100g
10g
Product Code. Quantity Particle Size unitSize
10608801 100 g 5 μm 100g
10618801 10 g 10 μm 10g
10618991 100 g 10 μm 100g
10388101 10 g 5 μm 10g
4 options
Product Code. 10608801
Brand: Macherey-Nagel™ 711360.100