missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfuric Acid, Supelco™

Product Code. 15846830
Change view
Click to view available options
Quantity:
2.5 kg
25 kg
Unit Size:
2.5kg
25kg
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15846830 25 kg 25kg
15836830 2.5 kg 2.5kg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15846830 Supplier Supelco Supplier No. 1002199025
 Restricted Item
Estimated Shipment: 09-11-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

EMPLURA™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Weight (g/mol) 97.09

Specifications

Melting Point 205°C (Decomposition)
Density 2.13 g/cm3 at 20°C
Quantity 25 kg
MDL Number MFCD00011603
Synonym Amidosulfuric acid, Amidosulfonic acid, Sulphamic acid, Sulfamidic acid, Amidosulfuric acid
Chemical Name or Material Amidosulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.