missing translation for 'onlineSavingsMsg'
Learn More

Amidosulfuric Acid, Supelco™

Product Code. 15823830
Change view
Click to view available options
Quantity:
100 g
250 g
Unit Size:
100g
250g
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
15823830 250 g 250g
15813830 100 g 100g
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
This item is not returnable. View return policy
Product Code. 15823830 Supplier Supelco Supplier No. 1001030250
 Restricted Item
Estimated Shipment: 14-01-2027
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For analysis EMSURE™

TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 5329-14-6
Molecular Weight (g/mol) 97.09

Specifications

Melting Point 205°C (Decomposition)
Density 2.13 g/cm3 at 20°C
Quantity 250 g
MDL Number MFCD00011603
Synonym Amidosulfuric acid, Amidosulfonic acid, Sulphamic acid, Sulfamidic acid, Amidosulfuric acid
Chemical Name or Material Amidosulfuric Acid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.