missing translation for 'onlineSavingsMsg'
Learn More
Learn More
AMICA/JAML Antibody (4E10) [Janelia Fluor™ 525], Novus Biologicals™
Armenian Hamster Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP1-43309JF525
This item is not returnable.
View return policy
Description
AMICA/JAML Monoclonal antibody specifically detects AMICA/JAML in Mouse samples. It is validated for Flow Cytometry,Immunoprecipitation
Specifications
| AMICA/JAML | |
| Monoclonal | |
| Janelia Fluor 525 | |
| adhesion molecule, interacts with CXADR antigen 1, AMICA, Gm638 | |
| Synthetic peptide corresponding to residues C(267) D G I L G D N F R P T Q(279) of mouse CARP. | |
| 0.1 mL | |
| Primary | |
| Mouse | |
| Purified |
| Flow Cytometry, Immunoprecipitation | |
| 4E10 | |
| 50mM Sodium Borate | |
| Armenian Hamster | |
| Protein A or G purified | |
| RUO | |
| 120425 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction