Learn More
AMG9810, MedChemExpress
Product Code. 16473538
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
Product Code. Quantity unitSize
16473538 10 mg 10mg
16459966 50 mg 50mg
16434666 100 mg 100mg
16406267 5 mg 5mg
4 options
Product Code. 16473538
Brand: Medchem Express HY101736
MedChemExpress AMG9810 is a selective and competitive vanilloid receptor 1 (TRPV1) antagonist with IC50 values of 24.5 and 85.6 nM for human and rat TRPV1, repectively.
AMG9810 is a selective and competitive vanilloid receptor 1 (TRPV1) antagonist with IC50 values of 24.5 and 85.6 nM for human and rat TRPV1, repectively.
Specifications
Chemical Name or Material AMG9810
CAS 545395-94-6
Color Off-White
Purity Grade Notes Research
Molecular Formula C21H23NO3
Quantity 10 mg
Solubility Information DMSO : ≥ 33 mg/mL (97.80 mM)
SMILES O=C(NC1=CC=C(OCCO2)C2=C1)/C=C/C3=CC=C(C(C)(C)C)C=C3
Molecular Weight (g/mol) 337.41
Formula Weight 337.41
Percent Purity 99.76%
Grade Research
Physical Form Solid
For Use With (Application) Neuroscience-Neuromodulation