missing translation for 'onlineSavingsMsg'
Learn More

AMG 837 calcium hydrate, MedChemExpress

Product Code. p-200096873
Encompass_Preferred
Click to view available options
Quantity:
5 mg
10 mg
50 mg
Unit Size:
10mg
50mg
5mg
This item is not returnable. View return policy

Product Code. 16488567

Brand: Medchem Express HY13967B

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

MedChemExpress AMG 837 calcium hydrate is a potent, orally bioavailable and partial agonist of GPR40/FFA1. AMG 837 calcium hydrate inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate could enhance insulin secretion and lower glucose levels in rodents.

AMG 837 calcium hydrate is a potent, orally bioavailable and partial agonist of GPR40/FFA1. AMG 837 calcium hydrate inhibits specific [3H]AMG 837 binding at the human FFA1 receptor with a pIC50 of 8.13. AMG 837 calcium hydrate could enhance insulin secretion and lower glucose levels in rodents.

Specifications

Chemical Name or Material AMG 837 calcium hydrate
CAS 1259389-38-2
Color White
Purity Grade Notes Research
Molecular Formula C26H21F3O3.1/2Ca.H2O
Quantity 5 mg
Solubility Information DMSO : ≥ 42 mg/mL (92.22 mM)
SMILES [O-]C(C[C@H](C#CC)C1=CC=C(C=C1)OCC2=CC=CC(C3=CC=C(C(F)(F)F)C=C3)=C2)=O.[0.5Ca2+].O
Molecular Weight (g/mol) 455.45
Formula Weight 455.45
Percent Purity 97.23%
Grade Research
Physical Form Solid
For Use With (Application) Metabolism-sugar/lipid metabolism
Show More Show Less

Research purposes only

Product Content Correction

The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.