Learn More
Amfenac Ethyl Ester, Mikromol™
Click to view available options
Quantity:
25 mg
Unit Size:
25mg
Description
Amfenac Ethyl Ester
Specifications
Specifications
| Chemical Name or Material | Amfenac Ethyl Ester |
| CAS | 61941-57-9 |
| Analyte or Component Names | Amfenac Ethyl Ester |
| Shipping Condition | Room Temperature |
| Molecular Formula | C17 H17 N O3 |
| Certifications/Compliance | ISO 17025 |
| Synonym | Amfenac Ethyl Ester, 2-Amino-3-benzoylbenzeneacetic acid ethyl ester, Ethyl (2-amino-3-benzoylphenyl)acetate, Ethyl 2-amino-3-benzoylbenzeneacetate |
| InChI Formula | InChI=1S/C17H17NO3/c1-2-21-15(19)11-13-9-6-10-14(16(13)18)17(20)12-7-4-3-5-8-12/h3-10H,2,11,18H2,1H3 |
| SMILES | CCOC(=O)Cc1cccc(C(=O)c2ccccc2)c1N |
| IUPAC Name | ethyl 2-(2-amino-3-benzoylphenyl)acetate |
| Show More |