Learn More
Ametryn, Dr. Ehrenstorfer
Specifications
Specifications
| Chemical Name or Material | Ametryn |
| CAS | 834-12-8 |
| Analyte or Component Names | Ametryn |
| Shipping Condition | +20°C |
| Molecular Formula | C9 H17 N5 S |
| Certifications/Compliance | ISO 17034 |
| InChI Formula | InChI=1S/C9H17N5S/c1-5-10-7-12-8(11-6(2)3)14-9(13-7)15-4/h6H,5H2,1-4H3,(H2,10,11,12,13,14) |
| SMILES | CCNc1nc(NC(C)C)nc(SC)n1 |
| IUPAC Name | 4-N-ethyl-6-methylsulfanyl-2-N-propan-2-yl-1,3,5-triazine-2,4-diamine |
| Molecular Weight (g/mol) | 227.33 |
| Show More |