Learn More
Cytiva Amersham™ Hyperfilm™ MP
Shop All Cytiva Products
Click to view available options
Dimensions (LxW):
5 x 7 in.
18 x 24 cm
8 x 10 in.
Quantity:
100 Sheets/Box, Single film bag
50 Sheets/Box, Indiv. film envelope
50 Sheets/Box, Single film bag
Unit Size:
100 sheets
50 sheets
Description
- High sensitivity for low concentration targets
- Optimized for direct and indirect autoradiography with 3H, 14C, 32P, 33P, 35S, 125I
- Clear background for excellent contrast and improved band visibility.
- Publication-quality images
- High resolution at an economical price
- Easy to use
Specifications
Specifications
| For Use With (Application) | 32P, 33P, 125I Southern, Northern, and Western blots. 32S, 33P, 35P sequencing. Fluorography of 3H, 14C, and 35S |
| Quantity | 50 Sheets/Box, Single film bag |
| Dimensions (LxW) | 5 x 7 in. |