Learn More
Cytiva Amersham™ Hyperfilm™ ECL
Product Code. 10479214 Shop All Cytiva Products
Change view
Click to view available options
Dimensions (LxW):
12.7 x 17.8 cm (5 x 7 in.)
18 x 24 cm (7.1 x 9.5 in.)
20.3 x 25.4 cm (8 x 10 in.)
Quantity:
100 Sheets/Box
50 Sheets/Box
Unit Size:
100 sheets
50 sheets
Product Code. Dimensions (LxW) Quantity unitSize
10479214 20.3 x 25.4 cm (8 x 10 in.) 50 Sheets/Box 50 sheets
10534205 18 x 24 cm (7.1 x 9.5 in.) 100 Sheets/Box 100 sheets
10752067 12.7 x 17.8 cm (5 x 7 in.) 50 Sheets/Box 50 sheets
10607665 18 x 24 cm (7.1 x 9.5 in.) 50 Sheets/Box 50 sheets
10075224 20.3 x 25.4 cm (8 x 10 in.) 100 Sheets/Box 100 sheets
5 options
Product Code. 10479214
Brand: Cytiva 28906838
Use with all blue and green light chemiluminescent systems. Cytiva Amersham™ Hyperfilm™ ECL is constructed from a clear base coated on both sides with a photographic emulsion that is protected by an anti-scratch layer.
  • Antistatic coating reduces risk of visible artifacts
  • Clear base offers high contrast for easy reading of images
  • Suitable for use with all blue and green light chemiluminescent systems
  • Film can be processed in automatic processors or manually using most common X-ray film developers and fixers
  • Chemiluminescent detection in Western, Southern, and Northern blots
TRUSTED_SUSTAINABILITY
Specifications
Product Type Amersham Hyperfilm ECL
For Use With (Application) Chemiluminescent detection in Western, Southern, and Northern blots.
Quantity 50 Sheets/Box
Storage Requirements 4°C
Dimensions (LxW) 20.3 x 25.4 cm (8 x 10 in.)