Learn More
Cytiva Amersham™ Hybond™ P Sandwich PVDF Membrane + 3MM Chr Paper
Shop All Cytiva Products
Click to view available options
Quantity:
10 + 20/Pk.
Unit Size:
Pack of 10
Specifications
Specifications
| Length (Metric) | 80 mm |
| Width (Metric) | 90 mm |
| Quantity | 10 + 20/Pk. |
| Pore Size | 0.45 μm |