Learn More
Cytiva Amersham™ ECL™ Western Blotting Detection Reagents
Product Code. 10340125 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
62.5 mL
125 mL
250 mL
375 mL
Sufficient For:
1000 cm2 Membrane
2000 cm2 Membrane
4000 cm2 Membrane
6000 cm2 Membrane
Unit Size:
Each
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Sufficient For unitSize
10340125 125 mL 2000 cm2 Membrane Each
10754557 62.5 mL 1000 cm2 Membrane Each
10773577 375 mL 6000 cm2 Membrane Each
10155854 250 mL 4000 cm2 Membrane Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Product Code. 10340125 Supplier Cytiva Supplier No. RPN2209
  • ONLINE EXCLUSIVE PRICE
    3310.00 NOK
    3127.00 NOK / Pack of 1
    Save 183.00 NOK
    5% Off
View all promotions
 Restricted Item
In Stock
Add to basket
Add to basket
Use for all routine Western blotting. Cytiva Life Sciences™ Amersham™ ECL™ Western Blotting Detection Reagents are the world's first commercially available chemiluminescent detection reagents and still one of the most widely used.
  • Detectable on Hyperfilm™ ECL or with CCD imager
  • Uses enhanced luminol-based detection which is suitable for all routine Western Blotting
Western Blotting
TRUSTED_SUSTAINABILITY
Specifications
Product Type ECL Western Blot Detection Reagent
For Use With (Application) Western Blot
Quantity 125 mL
Sensitivity ∼10 pg
Sufficient For 2000 cm2 Membrane
Kit Contents 125mL detection reagent, 1125mL detection reagent 2
Content And Storage 2°C to 8°C