Learn More
Cytiva Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System
Product Code. p-3676034 Shop All Cytiva Products
Change view
Click to view available options
Quantity:
For 2000 cm2 Membrane
For 4000 cm2 membrane
Labels 10 μg Nucleic Acid
Labels 5 μg Nucleic Acid
Product Type:
Detection Reagents
Hybridization Buffer
Labeling Module
Labeling and Detection System
Unit Size:
5µg
Each
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Product Type unitSize
10733797 Labels 5 μg Nucleic Acid Labeling Module 5µg
10195804 For 4000 cm2 membrane Detection Reagents Each
10554205 Labels 10 μg Nucleic Acid Labeling and Detection System Each
10411095 For 2000 cm2 Membrane Detection Reagents Each
10269004 Labels 5 μg Nucleic Acid Labeling and Detection System Each
10459774 For 4000 cm2 membrane Hybridization Buffer Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Product Code. 10733797 Supplier Cytiva Supplier No. RPN3005
Indeholder
45-000-922 and 45-000-923: Labeling reagent; Crosslinker; Control DNA; Blocking agent; ECL detection reagents and ECL Gold hybridization buffer
Use for fast and easy detection of medium-to-high-target amounts. Cytiva Life Sciences™ Amersham™ ECL™ Direct Nucleic Acid Labeling/Detection System are based on the direct labeling of DNA or RNA probes with horseradish peroxidase (HRP) in a simple 20 min chemical reaction
    • Direct probe labeling in a 10 min reaction, 1 h from hybridization to detection with ECL Direct, Hybond N+, and Hyperfilm ECL.
    • Eliminates handling, waste, and regulatory issues associated with the use of radioactivity.
    • No need to strip blots before reprobing.
    • For fast and easy detection of medium- to high-target amounts in applications such as colony/plaque screens, dot blots, and PCR product analyses.
    • Consistent results combining strong signals with very low backgrounds.
TRUSTED_SUSTAINABILITY
Tekniske data
Content And Storage 2°C to 8°C
For Use With (Application) Colony or plaque screens, dot blots and PCR product analyses
Includes DNA labelling reagent 0.5mL; Glutaraldehyde solution 0.5mL; Water 1.0mL; l Hind III control DNA (10μg/mL) 0.1 ml; Gold hybridization buffer 500mL; Blocking agent 25g
Product Type Labeling Module
Quantity Labels 5 μg Nucleic Acid
Test Time 20 min.
Signalord
  • Danger
Farekategori
  • Serious eye damage/eye irritation Category 2
  • Skin corrosion/irritation Category 2
  • Skin sensitiser Category 1
  • Respiratory sensitiser Category 1
  • Specific target organ toxicity Category 3
  • LONG-TERM AQUATIC HAZARD Chronic 3
Faresætning
  • H319-Causes serious eye irritation.
  • H315-Causes skin irritation.
  • H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
  • H317-May cause an allergic skin reaction.
  • H335-May cause respiratory irritation.
  • H412-Harmful to aquatic life with long lasting effects.
Sikkerhedssætning
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P273-Avoid release to the environment.
  • P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
  • P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
  • P405-Store locked up.
  • P501-Dispose of contents/container to .
Produkttitel
Vælg et problem
Ved at klikke på Send, anerkender du, at du kan blive kontaktet af Fisher Scientific med hensyn til den feedback, du har givet i denne formular. Vi deler ikke dine oplysninger til andre formål. Alle angivne kontaktoplysninger skal også vedligeholdes i overensstemmelse med vores Privatlivspolitik.