Learn More
Invitrogen™ RiboMinus™ Transcriptome Isolation Kit, bacteria
Product Code. 10063512
Click to view available options
Quantity:
12 preps
Unit Size:
1 set
Product Code. 10063512
Brand: Invitrogen™ K155004
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Includes
The RiboMinus Isolation Kits include RiboMinus Magnetic Beads, RiboMinus human/mouse, bacteria, or yeast probe, and Hybridization Buffer. The optional RiboMinus Concentration Module contains spin cartridges, wash tubes, recovery tubes, Binding Buffer, Wash Buffer, and RNase-free (DEPC-treated) water.
RiboMinus™ Transcriptome Isolation Kits are novel purification systems that enrich the whole spectrum of RNA transcripts by depleting large ribosomal RNA molecules in a total RNA sample.
RiboMinus™ Transcriptome Isolation Kits are novel purification systems that enrich the whole spectrum of RNA transcripts by depleting large ribosomal RNA molecules in a total RNA sample. These kits use a 5μ biotin-labeled, Locked Nucleic Acid (LNA) probe specific for large rRNA to remove 95 to 98% of rRNA molecules from human, mouse, bacteria, or yeast total RNA. This removal increases the representation of RNA transcript species, including non-poly(A) tailed, pre-processed RNA, small tRNAs, siRNA, snRNA, microRNA, fragmented mRNA, and other regulatory RNAs that cannot be purified via poly(A) selection methods.
Purified RNA is concentrated and suitable for numerous downstream applications, including microarray analysis, library construction, and qRT-PCR.
• Comprehensive analysis of the whole transcriptome without contamination or interference from rRNA
• More complete collection of transcribed elements of the genome—no reliance on poly(A) purification methods
• Cleaner, clearer microarray analysis due to higher signal-to-background ratios
• Not high-throughput compatible (Manual)
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage The RiboMinus™ Isolation Kits include RiboMinus™ Magnetic Beads, RiboMinus™ human/mouse, bacteria, or yeast probe, and Hybridization Buffer.
The optional RiboMinus™ Concentration Module contains spin cartridges, wash tubes, recovery tubes, Binding Buffer, Wash Buffer, and RNase-free (DEPC-treated) water.
Store the isolation modules at 4°C, and the concentration module at room temperature.
All reagents are guaranteed stable for 6 months when properly stored.
Isolation Technology Magnetic Bead
Purification Time 1.25 hr.
Sample Type Total RNA (Bacterial)
Final Product Type Transcriptome RNA
High-throughput Compatibility Not High-throughput Compatible (Manual)
For Use With (Application) Microarray Analysis, Real-Time Quantitative PCR (qPCR), Reverse Transcriptase PCR (RT-PCR), Northern Blotting, cDNA Library Construction
Product Line RiboMinus
Quantity 12 preps
Starting Material Amount Up to 10 μg total RNA
Target Transcriptome RNA
System Type RiboMinus
Yield 1 μg
Form Magnetic Bead