Learn More
Invitrogen™ Ambion™ mMESSAGE mMACHINE™ T7 Transcription Kit
Specifications
Specifications
| Concentration | 40 mM |
| For Use With (Application) | Gene Expression Analysis and Genotyping, in vitro Transcription |
| Includes | T7 Enzyme Mix, 10X Reaction Buffer, 2XNTP/CAP Solution, GTP Solution, pTRI-Xef, TURBO DNase, Ammonium Acetate Stop Solution, Lithium Chloride Precipitation Solution, Gel Loading Buffer II, and Nuclease-free Water |
| No. of Reactions | 25 |
| Product Type | Kit with manual |
| Quantity | 25 Reactions |