Learn More
Ambion™ Ambion™ In Vivo Pre-Designed siRNA
Description
Ambion In Vivo Predesigned siRNAs are specifically designed for gene silencing applications in animal models. These siRNAs are chemically modified to enhance stability, reduce immunogenicity, and improve delivery efficiency, making them excellent for in vivo studies. Ambion In Vivo Predesigned siRNAs allow researchers to investigate gene function in a physiological context, helping provide valuable insights into gene regulation mechanisms and disease processes. Built using the Silencer Select algorithm, these predesigned siRNAs help ensure high specificity and efficiency.
Ambion™ siRNAs are classic 21-mers which incorporate the latest improvements in siRNA design, off-target effect prediction algorithms, and chemistry. These siRNAs are guaranteed-to-silence based on their proven design.
RNA interference (RNAi) is the best way to effectively knock down gene expression to study protein function in a wide range of cell types. Use our GeneAssist™ Custom siRNA Builder to design and order a custom siRNA.
Specifications
Specifications
| Color | White |
| Content And Storage | Store in freezer (-5°C to -30°C). |
| Description | Ambion™ In-vivo Pre-designed siRNA, 5nmol, Powder, White, Standard Purity, Combined Knockdown Effectiveness and Stability, Easy Tracking of Administered siRNAs, Double-Stranded siRNAs That are Recognized by the RNA-induced Silencing Complex (RISC) to |
| RNAi Type | siRNA |
| Final Product Type | Ambion™ In Vivo Pre-Designed siRNA |
| Product Line | Ambion |
| Product Type | Ambion™ In-vivo Pre-designed siRNA |
| Purity | Standard |
| Quantity | 5 nmol |