Learn More
Invitrogen™ Ambion™ FirstChoice™ Human Brain Reference RNA, DNase-treated
Product Code. 10380345
Click to view available options
Quantity:
50 μg
Unit Size:
Each
Product Code. 10380345
Brand: Invitrogen™ AM6050
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Highest quality control and purity
Ambion FirstChoice™ Human Brain Reference RNA is a high-quality standard for human microarray analysis, pooled from multiple donors and several brain regions, thus ensuring unbiased coverage. Supplied in one tube containing 50μg. The Reference RNA passes the stringent QC criteria for purity and integrity that distinguishes all of FirstChoice RNA offerings and is the same RNA (identical manufacturing lot) used for the MicroArray Quality Control (MAQC) project.
DNA and RNA Purification and Analysis, RNA Extraction
Order Info
Shipping Condition: Dry ice
TRUSTED_SUSTAINABILITY
Specifications
Quantity 50 μg
For Research Use Only. Not for use in diagnostic procedures.