Learn More
Invitrogen™ Ambion™ FirstChoice™ Human Brain Reference RNA, DNase-treated
Description
Ambion FirstChoice™ Human Brain Reference RNA is a high-quality standard for human microarray analysis, pooled from multiple donors and several brain regions, thus ensuring unbiased coverage. Supplied in one tube containing 50μg. The Reference RNA passes the stringent QC criteria for purity and integrity that distinguishes all of FirstChoice RNA offerings and is the same RNA (identical manufacturing lot) used for the MicroArray Quality Control (MAQC) project.
DNA and RNA Purification and Analysis, RNA Extraction
Order Info
Shipping Condition: Dry ice
Specifications
Specifications
| Quantity | 50 μg |