missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Ambion™ ANTI-RNase (15-30U/μL)

Product Code. 10515395
Change view
Click to view available options
Quantity:
2500 U
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
10515395 2500 U Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 10515395 Supplier Invitrogen™ Supplier No. AM2690

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Specifications

Concentration 15 to 30U/μL
Content And Storage Store at -20°C. Do not store in a frost–free freezer or at -70°C.
For Use With (Application) Non-covalently binds and inactivates neutral pancreatic RNase A–type enzymes
Product Type ANTI-RNase
Quantity 2500 U
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.