Learn More
Amberlite™ IR-120, Cation Exchange Resin, for Ion Chromatography, gel type (hydrogen form, strongly acidic), Fisher Chemical™
Product Code. 10308063 Shop All Fisher Chemical Products
Change view
Click to view available options
Quantity:
100 g
500 g
Unit Size:
100g
500g
Product Code. Quantity unitSize
10308063 100 g 100g
10318063 500 g 500g
2 options
Product Code. 10308063
Brand: Fisher Chemical IE/1015/48
Request Bulk or Custom Format
Currently Unavailable: We have no information when this product will be available.